E-64
About this item
- Light Sensitive
CAS No.: 66701-25-5
Molecular Formula: C15H27N5O5
Formula Weight: 357.40
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in DMSO (25mg/ml), or water (20mg/ml)
Beilstein Reference: 1405664
Optical Rotation: +24° (c=1 in 1N HCl)
MDL No.: MFCD00080261
Selective and irreversible cysteine protease inhibitor
Specifications
- CAS No.:66701-25-5
- Storage conditions:Freezer
- MDL No.:MFCD00080261
- Protein synonyms:3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
- Protein/peptide name:E-64
- Molecular formula:C₁₅H₂₇N₅O₅
- Form:Powder
- Solubility:Soluble in DMSO (25mg/ml), or water (20mg/ml)
- Molecular weight:357.40
- Colour:White to off-white
Specifications
Related Information
Product Information
- Pack typePk
- J62933.MBGlass bottle for solids25 mg1 * 25 mg£403.00