2-Iodopropane for synthesis, Sigma-Aldrich®
8.43955.0250
: Merck
:
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
- Solubility:1.4 g/l (20°C)
- Melting Point:-90 °C
- Flash Point:42 °C
- Density:1.703 g/cm3 (25 °C)
:
:169.99 g/mol
:13,05234
:90 °C (1013 hPa)
:2,8 °C
:1,7 g/cm³ (20 °C)
:25 °C
:200-859-3
:2392
:3,III
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
:Ambient
:75-30-9
:C₃H₇I
:MFCD00001071