Northern Ireland
Order Entry
Masterflex® Ismatec® Microbore Tubing Fitting, size 12 (0.25mm ID) tubing, 316 SS, 2/pk
Masterflex® Ismatec® Microbore Tubing Fitting, size 12 (0.25mm ID) tubing, 316 SS, 2/pk
  MFLX34002-12
undefined
Masterflex® Ismatec® Microbore Tubing Fitting, size 12 (0.25mm ID) tubing, 316 SS, 2/pk
  MFLX34002-12
£97.20
  1 * 2 items
  • Barb size/tubing int.Ø (port 1):
    0.25 mm
  • Connection Type:
    Microbore
  • Flow pattern:
    Straight
  • Description:
    Microbore Tubing Fitting, size 12 (0.25mm ID) tubing, 316 SS, 2/pk
  • Environmentally Preferable:
  • Pk:
    2
  • Fitting material:
    Stainless steel, PETG