Masterflex® L/S® Ultrasonic Flow Sensor for L/S® 17 Tubing
MFLX33501-17
CE Compliant
IP65 washdown
- Catalog No:
- MFLX33501-17
- MFLX33501-82
- MFLX33501-15
- MFLX33501-24
- MFLX33501-36
- MFLX33501-73
- MFLX33501-16
- MFLX33501-25
- MFLX33501-91
- Accuracy:
- 2 to 4% after using calibration software
- 2 to 4% after using calibration software
- 2 to 4% after using calibration software
- 2 to 4% after using calibration software
- 2 to 4% after using calibration software
- 2 to 4% after using calibration software
- 2 to 4% after using calibration software
- 2 to 4% after using calibration software
- 2 to 4% after using calibration software
- Max. operating temperature:
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- 60 °C (140 °F)
- Enclosure rating:
- IP65
- IP65
- IP65
- IP65
- IP65
- IP65
- IP65
- IP65
- IP65
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- Maximum Flow Rate:1700 ml/min (26,95 gal./h)
- Min. flow rate:30 ml/min (0,48 gal./h)
- Output:RS-485, 4-20 ma, 0-20 kHz, or PNP-NPN
- Accuracy:2 to 4% after using calibration software
- Description:Ultrasonic Flow Sensor for L/S® 17 Tubing
- Seq:03
- Environmentally Preferable:
- Enclosure rating:IP65
- Max. operating temperature:60 °C (140 °F)
- Warranty:1 year
- Pk:1
- Housing material:Aluminium