Barium chloride, Suprapur® for trace analysis, Supelco®
: Merck
:10361-37-2
:
:BaCl₂
:MFCD00003445
:208.24 g/mol
:12,00998
:1560 °C (1013 hPa)
:∼963 °C
:2,33 g/cm³ (20 °C)
:1564
:6.1,III
:M-Clarity™ quality level = MQ100
:233-788-1
:GHS06
:Ambient
- Solubility:375 g/l (20 °C)
- Melting Point:963 °C
- Molar Mass:208.23 g/mol
- Bulk Density:1350 kg/m3
- Boiling Point:1560 °C
- Density:3.9 g/cm3 (20 °C)
- pH:5 - 8 (50 g/l, H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
- Pack typePk
- 1.01716.0500Plastic bottle500 g1 * 500 g£1,160.00