Would you like to log in again?
Laboratory bottles, wide neck, with GL80 thread
Supplier: VWR Collection
Borosilicate glass, clear or amber with screw caps and pouring rings
The large opening can accommodate even large volume funnels and gives easy access to scoops or spatulas. Amber to protect light-sensitive materials.
  • Very good chemical resistance, high temperature resistance
  • Extra wide opening allows easy cleaning
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Clear glass
Capacity Pk VWR Catalog Number Quantity
500 ml 1 215-3743
1000 ml 1 215-3744
2000 ml 1 215-3745 Unavailable
5000 ml 1 215-3746
Amber glass
Capacity Pk VWR Catalog Number Quantity
500 ml 1 215-3747
1000 ml 1 215-3748
2000 ml 1 215-3749
5000 ml 1 215-3773
unavailable This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty