Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Buffer solutions, 20 °C AVS TITRINORM®

Supplier: VWR Chemicals

  • Solutions prepared from AnalaR® NORMAPUR® grade analytical reagents
  • True analytical value on the label to an accuracy of ± 0,02 pH units (pH 1- 11) (± 0,05 for pH 12.00 buffer)
  • Traceable to SRM from NIST
  • Batch number and packing date for traceability
  • Certificate of analysis available from www.vwr.com
Ordering information: If significantly large volumes are required please contact your local VWR sales office for a customised offer.

Analysed by an independent accredited control laboratory ISO EN 17025

For full information concerning health and safety data please see the vwr.com website.

Packaging information: * 5 litre plastic bottle; ** Bag in a box 5 or 10 litre; *** 1 litre dosing bottles



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now

Buffer solutions in bottles
pH value pH accuracy pH buffer type Reference temperature Pack type Pk VWR Catalog Number Quantity
9,00 ±0,02 Borate 20 °C Bag-in-box (Cubitainer) 5 l 32039.374
9,00 (blue coloured)*** ±0,02 Borate 20 °C Dosing Bottle (Plastic) 1 l 32046.290
1,00 ±0,02 Glycine 20 °C Plastic bottle 1 l 32031.297
Get a discount if you order 6 or more. Login in now for details.
2,00 ±0,02 Citrate 20 °C Plastic bottle 500 ml 32032.260
Get a discount if you order 6 or more. Login in now for details.
2,00 ±0,02 Citrate 20 °C Plastic bottle 1 l 32032.291
Get a discount if you order 6 or more. Login in now for details.
3,00 ±0,02 Citrate 20 °C Plastic bottle 1 l 32033.294
Get a discount if you order 6 or more. Login in now for details.
3,00 ±0,02 Citrate 20 °C Bag-in-box (Cubitainer) 5 l 32033.374
4,00 ±0,02 Phthalate 20 °C Plastic bottle 500 ml 32095.264
Get a discount if you order 6 or more. Login in now for details.
4,00 ±0,02 Phthalate 20 °C Plastic bottle 1 l 32095.297
Get a discount if you order 6 or more. Login in now for details.
4,00* ±0,02 Phthalate 20 °C Plastic bottle 5 l 32095.366
Get a discount if you order 4 or more. Login in now for details.
4,00** ±0,02 Phthalate 20 °C Bag-in-box (Cubitainer) 5 l 32095.377
4,00 (red coloured) ±0,02 Phthalate 20 °C Plastic bottle 500 ml 32044.268
Get a discount if you order 6 or more. Login in now for details.
4,00 (red coloured)*** ±0,02 Phthalate 20 °C Dosing bottle (Plastic) 1 l 32044.290
4,00 (red coloured) ±0,02 Phthalate 20 °C Plastic bottle 1 l 32044.292
Get a discount if you order 6 or more. Login in now for details.
4,00 (red coloured)** ±0,02 Phthalate 20 °C Bag-in-box (Cubitainer) 5 l 32044.372
4,00 (red coloured)** ±0,02 Phthalate 20 °C Bag-in-box (Cubitainer) 10 l 32044.418
4,66 ±0,02 Acetate 20 °C Plastic bottle 500 ml 98192.260
Get a discount if you order 6 or more. Login in now for details.
4,66 ±0,02 Acetate 20 °C Plastic bottle 1 l 98192.290
Get a discount if you order 6 or more. Login in now for details.
5,00 ±0,02 Citrate 20 °C Plastic bottle 1 l 32035.291
Get a discount if you order 6 or more. Login in now for details.
6,00 ±0,02 Citrate 20 °C Plastic bottle 1 l 32036.294
Get a discount if you order 6 or more. Login in now for details.
6,88 ±0,02 Phosphate 20 °C Plastic bottle 500 ml 83601.260
Get a discount if you order 6 or more. Login in now for details.
6,88 ±0,02 Phosphate 20 °C Plastic bottle 1 l 83601.290
Get a discount if you order 6 or more. Login in now for details.
7,00 ±0,02 Phosphate 20 °C Plastic bottle 500 ml 32096.267
Get a discount if you order 6 or more. Login in now for details.
7,00 ±0,02 Phosphate 20 °C Plastic bottle 1 l 32096.291
Get a discount if you order 6 or more. Login in now for details.
7,00* ±0,02 Phosphate 20 °C Plastic bottle 5 l 32096.360
Get a discount if you order 4 or more. Login in now for details.
7,00** ±0,02 Phosphate 20 °C Bag-in-box (Cubitainer) 5 l 32096.371
7,00 ±0,02 Phosphate 20 °C Bag-in-box (Cubitainer) 10 l 32096.400
7,00 (green coloured) ±0,02 Phthalate-phosphate 20 °C Plastic bottle 500 ml 32045.262
Get a discount if you order 6 or more. Login in now for details.
7,00 (green coloured)*** ±0,02 Phthalate-phosphate 20 °C Dosing Bottle (Plastic) 1 l 32045.290
7,00 (green coloured) ±0,02 Phthalate-phosphate 20 °C Plastic bottle 1 l 32045.295
Get a discount if you order 6 or more. Login in now for details.
7,00 (green coloured) ±0,02 Phthalate-phosphate 20 °C Bag-in-box (Cubitainer) 5 l 32045.375
7,00 (green coloured) ±0,02 Phthalate-phosphate 20 °C Bag-in-box (Cubitainer) 10 l 32045.412
7,00 (green coloured) ±0,02 Phthalate-Phosphate 20 °C Bag-in-box (Cubitainer) 10 l 30926.412 Unavailable
8,00 ±0,02 Borate 20 °C Plastic bottle 1 l 32038.291
Get a discount if you order 6 or more. Login in now for details.
9,00 ±0,02 Borate 20 °C Plastic bottle 500 ml 32039.261
Get a discount if you order 6 or more. Login in now for details.
9,00 ±0,02 Borate 20 °C Plastic bottle 1 l 32039.294
Get a discount if you order 6 or more. Login in now for details.
9,00 (blue coloured) ±0,02 Borate 20 °C Plastic bottle 500 ml 32046.265
Get a discount if you order 6 or more. Login in now for details.
9,00 (blue coloured) ±0,02 Borate 20 °C Plastic bottle 1 l 32046.298
Get a discount if you order 6 or more. Login in now for details.
9,00 (blue coloured)** ±0,02 Borate 20 °C Bag-in-box (Cubitainer) 5 l 32046.378
9,22 ±0,02 Borate 20 °C Plastic bottle 500 ml 32097.261
Get a discount if you order 6 or more. Login in now for details.
9,22 ±0,02 Borate 20 °C Plastic bottle 1 l 32097.290
Get a discount if you order 6 or more. Login in now for details.
10,00 ±0,02 Borate 20 °C Plastic bottle 500 ml 32040.260
10,00 ±0,02 Borate 20 °C Plastic bottle 1 l 32040.298
Get a discount if you order 6 or more. Login in now for details.
10,00 ±0,02 Borate 20 °C Bag-in-box (Cubitainer) 5 l 32040.378
10,00 (total hardness in water) ±0,05 Ammonia 20 °C Plastic bottle 500 ml 98213.260
Get a discount if you order 6 or more. Login in now for details.
10,00 (yellow) ±0,02 Borate 20 °C Plastic bottle 500 ml 85680.260
10,00 (yellow) ±0,02 Borate 20 °C Plastic bottle 1 l 85680.290
10,00 (yellow) ±0,02 Borate 20 °C Plastic bottle 1 l 85680.295
11,00 ±0,05 Phosphate 20 °C Plastic bottle 1 l 32041.292
Get a discount if you order 6 or more. Login in now for details.
12,00 ±0,05 Phosphate 20 °C Plastic bottle 1 l 32042.295
Get a discount if you order 6 or more. Login in now for details.

Buffer solution sets
pH value Colour Reference temperature Pack type Pk VWR Catalog Number Quantity
10,00/7,00/4,00 Blue/green/red 20 °C Plastic bottle (3×100 ml) 1 83610.600
10,00/7,00/4,00 Blue/green/red 20 °C Plastic bottle (3×250 ml) 1 83610.610

Production
pH value pH accuracy pH buffer type Reference temperature Pack type Pk VWR Catalog Number Quantity
4,00 (red coloured) ±0,02 Phthalate 20 °C Plastic bottle 1 SET VWRC32044.292BOM1
4,00 (red coloured) ±0,02 Phthalate 20 °C Plastic bottle 1 SET VWRC32044.292BOM2
4,00 (red coloured)** ±0,02 Phthalate 20 °C Bag-in-box (Cubitainer) 1 SET VWRC32044.372BOM
4,00* ±0,02 Phthalate 20 °C Plastic bottle 1 SET VWRC32095.366BOM
unavailable This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty