Disinfection Control
Qualitative and quantitative test strips for checking cleaning and disinfection processes in food and related industries.
Merckoquant (MQ) is a range of qualitative strips using a colour scale providing an indication of a range of concentration of a parameter within seconds with no need for auxillary reagents.
Reflectoquant (RQ) offers quantitative strips that need to be measured in a small RQflex® 10 reflectometer providing a result in less than a minute.
Brochures to order: w286183 " Mission Accomplished from Merck" and w285102 "Water and food analytics"