Media fill culture media
Culture media for media fill tests. Granulated or ready to use, for the pharmaceutical or food and beverage market.
OTHER MICROBIOLOGY SOLUTIONS
TSB for the pharmaceutical industry
GMP requires pharmaceutical companies to test their aseptic production process for microbiological contamination, the so called 'media fill test'
Linden grain medium for low acid aseptic beverage production
Linden Grain medium is specifically designed to test low-acid aspetic beverage production lines.