Just PROVE it
Spectroquant® Prove - simplify water analysis with a new class of spectrophotometer
New range of instruments designed to meet all your requirements for water analysis - wastewater, process water or drinking water and beverages.
Request Demo/Quote- Live ID, a new, data-rich 2-D barcode on all cell and reagent tests for recognising methods and transferring ‘live’ data like expiry, lot number together with automatic, complementary calibration updates
- Cell Test Port - direct insertion of round cell tests without opening any lid
- AQA prime - complete verification and documentation for all stages of quality control
- Easy cleaning – removable cell holder
- Matrix Check easily detects interferences
Spectroquant® PROVE 100 | Spectroquant® PROVE 300 | Spectroquant® PROVE 600 |
![]() |
![]() |
![]() |
Routine applications | Sensitive measurements | Complex analysis |
The best choice for use with cell tests and Visible measurements |
Long lasting Xenon lamp makes it suitable for intensive use in UV/Visible |
High-end UV/Visible optics and cuvettes up to 100 mm. Excellent resolution and sensitivity |
1.73016.0001 | 1.73017.0001 | 1.73018.0001 |
Introduction
How to start the Prove
How to navigate through the functions
How to use Prove with the Spectroquant® cell test kits and reagent sets
How to easily clean the Prove
How to select the language, date and time
Datatransfer - Data on demand - easy to filter - easy to share
How to check interfering matrix
Be sure of your system: Use AQA Analytical Quality Assurance