Digital Intelligent Measurement for more Accuracy and Flexibility!
Initially pH measurement started with a reference electrode and a measuring cell. In time, the combined electrode took over, followed by the combined electrode with built-in temperature sensor. Today, we take another step into the future with the innovative Digital sensors for pH, redox, conductivity, dissolved oxygen and even turbidity.
Digital Intelligent sensors are:
- More accurate as the signal is converted directly in the electrode
- Intelligent as serial numbers and settings including calibration dates are transferred automatically to the meter for best data management ever
- Easy to use as each unique sensor is recognised by the meter
and much more ....
VWR® Multiparameter Digital MD 8000
Advantages of a Digital measurement system
Watch this video to see how these innovations make your work easier, more accurate and give you flexibility for future projects!
Production Selection
pHenomenal MD 8000 L
One channel bench meter
6 parameters: pH/temperature, redox, conductivity, TDS, Salinity, Dissolved oxygen and turbidity.
Memory 5000 Datasets GLP
pHenomenal MD 8000 H
One channel portable meter
6 parameters: pH/temperature, redox, conductivity, TDS, Salinity, Dissolved oxygen and turbidity
Memory 5000 Datasets GLP
WTW® Multiparameter Digital IDS systems
Product Selection
WTW® IDS Multiparameter Lab meters
1 to 3 channel bench meter
6 parameters: pH/temperature, redox, conductivity, TDS, Salinity, Dissolved oxygen and turbidity.
Memory 5000 Datasets GLP
WTW® IDS Multiparameter Portable Meters
1 to 3 channel portable meter
6 parameters: pH/temperature, redox, conductivity, TDS, Salinity, Dissolved oxygen and turbidity
Memory 5000 Datasets GLP
HACH® Mono- & Multi-parameter HQD systems
Hach HQd IntelliCALâ„¢ durable probes
HQD meter, Have a look at Hach's HQd IntelliCAL durable water testing probes.
Hach® HQD Mono- and multi-parameter lab meters
1- to 2-channel meter: pH/temperature, redox, conductivity, TDS, salinity, dissolved oxygen and ISE. 500 data set GLP memory.
Hach® HQD mono- and multi-parameter portable meters
1- to 2-channel meter: pH/temperature, redox, conductivity, TDS, salinity, dissolved oxygen and ISE. 500 data set GLP memory.