Quantus Under Garments
In order to optimize the Quantus® garment system, we recommend the use of abrasion-proof, functional under garments (Underwear).
High quality blends in polyester, microfibre or cotton/polyester that minimize loose fibers are available. Additional comfort is achieved through optimal moisture and heat transport.
Besides physiological aspects and comfort, aspects of hard-wearing work clothes as well as CI-requirements and modern styles and colors are becoming more and more important when choosing under garments.
The product range of Quantus Under Garments includes:
  • Sweatshirts (long sleeves) 
  • T-Shirts (short sleeves)
  • Trousers
  • Jackets
  • Socks
Fabrics for Quantus Under Garments 
Model Overview Quantus Under Garments
Sizing for Quantus Under Garments
Underwear Standard
Sweatshirt (long sleeves with knitted kuffs), T-Shirts (short sleeves), Trousers (with rubber band at ankle height) available. Made of microfibre, antimicrobial finish
AreaDress
Top and Trousers. antistatic and antimicrobial equipped. Keenly softer feeling, with tightly texture tie and excellent breathability.
T-Shirt
Functional underwear (Jersey) with short sleeves and bordered cuffs.
Socks
Scks with low abrasion and washable to 30°C.