Tyvek Packaging
Features:
  • Composite of polyethylene foil and uncoated Tyvek®
  • Transparent foil offers the advantage of product visibility
  • Steam-sterilisable up to 125°C
  • Ethylene oxide- and gamma sterilisable
  • Very low gas residue (for instance ETO) gas sterilisation shortens the airing out time
  • Sterilisation indicators on the packaging
  • Extremely low particle contamination
  • Low fluff
  • Outstanding barrier characteristics
  • High-quality sealing and burst strength
  • Easily heat-sealed (sealing temperature up to max. 120°C)
  • Foils are suitable for printing
  • 100% recyclable
  • Optionally available as flat bag or easy-tear-bag, to minimize particle contamination
  • Available in various cleanroom classes
Flat bag:
  • Various widths and lengths
  • Standard thickness: 0.100 mm
  • Available with or without side pleat certificate of conformity with each delivery
Tube:
  • Various widths
  • Length: 100 m
  • The ideal packaging for oversized sterilized goods
Product selection
Tyvek-Packaging
Please choose from a list of products and refine your search by attributes...